sPLA2 Activity Kit, Enzo Life Sciences
# de catalogue 89141-664
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Sensitivity:3.85 Units/mL (range 5 - 80 Units/mL)
- No. of wells:96 Wells
- Cat. no.:89141-664
- Supplier No.:907-002
Spécifications
A propos de cet article
For the quantitative determination of sPLA2 in culture supernatants, cell lysates, and buffers from any species.
The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour.
Ordering information: Contains Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent.
Customers Who Bought This Also Bought