Samco™ Round Base Test Tubes, 12×75 mm, Thermo Scientific
Fournisseur: Samco
Certificats
A propos de cet article
Samco™ round based 12×75 mm tubes are suitable for most laboratory applications.
- Available with or without cap
Available in clear polypropylene for chemical resistance and strength, or polystyrene for superior clarity.
Ordering information: Ambient storage and shipping recommended for tubes.
Packaging: Choice of packaging formats: sterile (bulk or individually wrapped) or non sterile (bulk or blister tray).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.