About this item
Prevent bio-hazardous spills while conserving laboratory space with stackable Samco™ Bio-Tite™ Narrow-Mouth 90mL (3oz.) 48mm Specimen Containers
- Plug seal cap ensures excellent leak-resiatant performance
- Cap and base knurls ensure ease of handling
- Containers are stackable on top of each other for ease of storage
- Supplied nonsterile or gamma irradiated for guaranteed sterility
These shatter-resistant containers feature a leak-resistant seal and are ideal for the collection and transport of both liquid and solid samples. Available in both sterile and nonsterile formats, with a range of cap colors and label types to suit most applications.