- Size:1 Library (500 µl/well)
- Cat. no.:89156-266
- Supplier No.:2810-0500
The Screen-Well neurotransmitter library contains 681 CNS receptor ligands in a 96-well format.
Includes 13 classes of receptor ligands in 10 deep-well plates (Adrenergis; Histaminergics, Melatonin ligands; Dopaminergics; Ionotropic; Glutamatergics; Serotonergics; Metabotropic; Glutamatergics; Opioids, Sigma ligands; GABAergics; Cholinergics; Purinergics; Adenosines). Each ligand is supplied in solution in a biocompatible solvent at a standard concentration of 10mM (Opioid peptides are supplied at a concentrations of 1mM). Plates are available as a complete set, or individually. Supporting data are included.
Ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays, and for routine pharmacological applications.