Shipper, Multisample EAS Ambient, Therapak®
# de catalogue 10029-416
Fournisseur: Therapak, LLC
Spécifications
- Description:Large transport box, multi-sample shipper
- Cat. no.:10029-416
- Supplier No.:56883
Spécifications
A propos de cet article
Large capacity ambient shipping system accommodates multiple specimen transport bags for consolidation at collection site and shipping to testing laboratory
Features a pop-up style box that is easy to assemble. Easy-fold flaps on both ends of the box enable the shipper to easily slide the included specimen bag and absorbent containing sample tubes inside the box prior to securing the flaps for shipment. Includes an Exempt Animal Specimen label and biohazard symbol label.
System includes a large transport box (ID: 11 x 10.5 x 4"; OD: 11.25 x 10.75 x 4.25") with easy lock flaps on both ends of the box, a 13 x 18" biohazard transport bag, and 12 x 12" absorbent sheet. Holds up to ten 6 x 9" specimen transport bags.
Customers Who Bought This Also Bought