Shipper, EHS Refrigerated, Therapak®
Catalog # 10029-402
Supplier: Therapak, LLC
Specifications
- Description:EHS refrigerated shipping system
- Cat. no.:10029-402
- Interior Dimensions:8×6×8”
- Exterior Dimensions:11×9×11”
- Supplier No.:56876
Specifications
About this item
EHS shipping systems for refrigerated specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and box (OD: 11 x 9 x 11”), which is ideal for overnight shipments via common carriers such as DHL and FedEx
Refrigerated shipping system includes: 8-oz cold packs, specimen bag, absorbent material, Exempt Human Specimen label, biohazard label and cushioning material. Each case includes one set of packing instructions.
Customers Who Bought This Also Bought