Shipper, NanoCool, Refrigerated 2...8 °C, Therapak®
Fournisseur: Therapak, LLC
Certificats
A propos de cet article
Therapak®’s NanoCool 2-8°C Specimen Shipping System features a self-cooling system built into the lid.
Temperature performance testing shows the system maintains 2-8°C when tested against a ISTA-7D 48-hour summer profile. The NanoCool’s durable design also passes drop testing to a modified ISTA-3A sequence.
With no refrigeration required, activate the cooling process with the simple push of a button and your contents will remain in the 2° to 8°C range for up to 48 hours with no further conditioning or temperature control necessary.
Customers Who Bought This Also Bought