Shipper, Category B Refrigerated, 2...8 °C, Therapak®
# de catalogue 10029-326
Fournisseur: Therapak, LLC
Spécifications
- Description:Cat B refrigerated, 2-8°C
- Cat. no.:10029-326
- Exterior Dimensions:12×12×12"
- Supplier No.:38028
Spécifications
A propos de cet article
Refrigerated Shipping System has been field tested for 2 - 8° C temperature maintenance for 48 hours
System includes: 12 x 12 x 12” 200# corrugated outer box, EPS cooler with 21/2” all around wall thickness, ambient gel wrap, 95kPa specimen transport bag with absorbent to hold up to six specimen tubes, refrigerant packs, list of contents card, filler material, and required labels for IATA PI650 marking and compliance. Packing Instructions are included to facilitate use for safe and compliant shipping.
Customers Who Bought This Also Bought