VWR® Strip Washblocker
# de catalogue 10147-836
Fournisseur: VWR International
Spécifications
- Description:Strip Washblocker
- Cat. no.:10147-836
Spécifications
A propos de cet article
Use this low-profile spill dike for routine containment of washdown and sanitizing liquid.
- Tacky urethane construction
- Tear-resistant polypropylene top
- Low-profile design
- Reusable
- Highly visible color
Block doorways, protect carpeting, and channel liquid to drains.
Tacky urethane construction creates a leakproof seal on dry floors. Tear-resistant polypropylene top withstands repeated use.
Lightweight strip is easy to handle and store. Reusable barrier cleans up with soap and water. Bright-green color is highly visible.
Ordering information: For smooth surfaces.
Customers Who Bought This Also Bought