Samco™ Bio-Tite™ Midstream Urine Collection Kit, Thermo Scientific
Fournisseur: Samco
Certificats
A propos de cet article
Collect urine samples using the Samco™ Bio-Tite™ Midstream Urine Collection Kit
- Easy grip top and bottom knurls for ease of handling
- Available in two convenient formats: with or without funnel
- Instructions for use and wet wipes included in every kit
Bio-Tite™ closure included for assured leak-resistant performance.
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for containers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.