ContactUsLinkComponent
Shipper, Category B Frozen, 2" Cryogenic Box, Therapak®
Shipper, Category B Frozen, 2" Cryogenic Box, Therapak®
Catalog # 10029-304
Supplier:  Therapak, LLC
IATA-B Frozen Shipper, 4/Cs
Shipper, Category B Frozen, 2" Cryogenic Box, Therapak®
Catalog # 10029-304
Supplier:  Therapak, LLC
Specifications
  • Description:
    Cat B frozen, 2" cryogenic box
  • Cat. no.:
    10029-304
  • Supplier No.:
    37818
Specifications
About this item
Frozen specimen shipping system includes: transport box (OD: 15x13x12"), medium cooler (ID: 12x10x9"), 2" cryogenic vial box, absorbent pad, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance
Holds up to eighty-one cryogenic vials (up to 2 mL) for overnight shipments. System requires use of 95kPa compliant cryogenic vials.
Customers Who Bought This Also Bought