Shipper, Category B Frozen, Large Canister, Therapak®
# de catalogue 10029-302
Fournisseur: Therapak, LLC
Spécifications
- Description:Cat B frozen, Large canister
- Cat. no.:10029-302
- Supplier No.:37817
Spécifications
A propos de cet article
Frozen specimen shipping system.
- Holds up to eighteen tubes for overnight shipments
Includes transport box (OD: 15x13x12"), medium cooler (ID: 12x10x9"), large 95kPa canister, three Aqui-Pak™ segmented absorbent pouches, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance.
Customers Who Bought This Also Bought