Order Entry
ContactUsLinkComponent
Shipper, Category B Refrigerated, 95 kPa Bags, 18-Tube, Therapak®
Shipper, Category B Refrigerated, 95 kPa Bags, 18-Tube, Therapak®
Catalog # 10029-300
Supplier:  Therapak, LLC
IATA-B-Refrigerated Shipper, 4/Cs
Shipper, Category B Refrigerated, 95 kPa Bags, 18-Tube, Therapak®
Catalog # 10029-300
Supplier:  Therapak, LLC

Specifications

  • Description:
    Cat B refrigerated, 95kpa bags, 18-tube
  • Cat. no.:
    10029-300
  • Supplier No.:
    37816

Specifications

About this item

Refrigerated Shipping System includes transport box (OD: 15x13x12"), medium cooler (ID: 12x10x9"), three 95kPa bags, three Aqui-Pak™ segmented absorbent pouches, 8 oz

refrigerant packs, cushioning material, list of contents card, and required labels for IATA PI650 marking and compliance. Holds up to eighteen tubes for overnight shipments.

Customers Who Bought This Also Bought