Shipper, Category B Refrigerated, 95 kPa Bag, 6-Tube, Therapak®
# de catalogue 10029-294
Fournisseur: Therapak, LLC
Spécifications
- Description:Cat B refrigerated, 95kpa bag, 6-tube
- Cat. no.:10029-294
- Supplier No.:37813
Spécifications
A propos de cet article
Refrigerated Shipping System includes: transport box (OD: 11x9x11"), small cooler (ID: 8x6x8"), 95kPa bag, Aqui-Pak™ segmented absorbent pouch, 8 oz
refrigerant packs, cushioning material, list of contents card, and required labels for IATA PI650 marking and compliance. Holds up to six tubes for overnight shipments.
Customers Who Bought This Also Bought