Shipper, Category B Frozen, Medium Canister, Therapak®
Catalog # 10029-262
Supplier: Therapak, LLC
Specifications
- Description:Cat B frozen, Medium canister
- Cat. no.:10029-262
- Supplier No.:37806
Specifications
About this item
Frozen specimen shipping system includes: transport box (OD: 11x9x11"), small cooler (ID: 8x6x8"), medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance
Holds up to six tubes for overnight shipments.
Customers Who Bought This Also Bought