Mini Balance Bank, Small Workstation for Organizing Weighing Supplies, White PVC, TrippNT
# de catalogue 11376-007
Fournisseur: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Color:White
- Material:PVC
- W×D×H:30.5 x 12.7 x 30.5 cm (12 x 5 x 12")
- Supplier No.:50077
Spécifications
A propos de cet article
TrippNT's Mini Balance Bank Workstation is designed to hold standard weight papers, cups and tools. This organizer can also be used a a personal workstation or dedicated instrument organizer.
- White PVC construction
- 3 shelves, 6 slot tool organizer, 8 compartments
- Built-in Kimwipe dispenser and tool holder (for small tools)
- Keeps supplies clean, dry, and neat
- Simple to use and easy to fill
- Useful around scales, balances, pH meters
- American made by TrippNT
- Total replacement lifetime guarantee
The built-in tool holder and Kimwipe dispenser make it easy to grab a tool or tissue. Made from bright white PVC this workstation can be placed on countertop or a bench work surface. Ships fully assembled.
Weight: 1.36 kg (3 lbs)
Customers Who Bought This Also Bought