Samco™ Round Base Test Tubes, 17x100mm, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Samco™ round based 17×100 mm tubes are suitable for most laboratory applications.
- Choice of packaging formats: sterile or nonsterile
- Available with or without cap
- Polypropylene or polystyrene construction
Available in clear polypropylene for chemical resistance and strength, or polystyrene for superior clarity.
Ordering information: Ambient storage and shipping recommended for tubes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.