About this item
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ Wide-Mouth 40mL (1oz.) 48mm Specimen Containers
- Plug seal cap ensures excellent leak-resiatant performance
- Cap and base knurls ensure ease of handling
- Supplied nonsterile
These stackable, shatter-resistant containers are manufactured from virgin polypropylene and are ideal for the collection and transportation of both liquid and solid samples. Supplied bulk packed, containers are ideal for pre-fill applications.