A propos de cet article
Chewable, flavored, medicated 5 g tablet replaces more stressful dosing methods
Base Meal Ingredients: Sucrose, Dextrose, Mineral Oil, Artificial and Natural Flavors, Calcium Silicate, Magnesium Stearate.
Note: A signed MD Form is required for all Medications except Aspirin and L-Tryptophan. Please Fax the completed form to 908-284-4753. The form will remain on file for one year. Refills are permitted as long as the prescribing veterinarian listed on the request form is still affiliated with the requesting institution.
Tablet consistency ensures accurate dosing; scored for flexible dose administration. Cold tableting process eliminates drug degradation. No heat or water used in processing. Highly palatable sugar-based formula with excellent drug-masking capabilities. Reduces time, labor and stress compared to traditional dosing methods. Medications can be customized to meet specific requirements. Shelf Life: 6-24 months as dictated by the drug expiration date.