Drum Labels, Brady
# de catalogue CA12000-886
Fournisseur: Brady Worldwide
Spécifications
- Color:Blue on White
- Description:Notice drum labels
- Material:Paper
- Thickness:0.0038"
- Cat. no.:CA12000-886
- Legend:This Container Has Been Triple Rinsed
- Dimensions:152.4×152.4 mm (6×6")
- Supplier No.:60364
Spécifications
A propos de cet article
These durable labels are used to easily identify and sort waste for disposal.
- Labels remove cleanly, leaving no residue on surface
Made of coated Paper with pressure sensitive acrylic adhesive which can adheres to most surfaces.
Certifications: Meet D.O.T. requirements HM-181 regulation.
Customers Who Bought This Also Bought