A propos de cet article
Active universal cleaning agent for the main wash cycle, which cleans even heavily soiled items and removes dried or burned-on residues. Particularly suitable for the removal of starch and protein residues. Extran® AP 12 alkaline powder does not foam even during heavy agitation of the solution in a washing machine.
- Free from organic surfactants and emulsifiers
- Powder form
- Alkaline
- Surfactant-free and chlorine-free
- Free from odorants/dyestuffs
Dosing
The ideal dosing depends on the hardness of the water and the level of contamination of the item to be cleaned.
Under normal conditions, the concentration for application is 0.2–0.4%, i.e. 20–40 g of Extran® AP 12 are used for around 10 L of water
pH value (1%; water) ≥ 12.5
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- CA1.07563.9025
- CA1.07563.2000
- CA1.07563.9010
- Formulation:
- Phosphates, sodium hydroxide, alkali salts
- Phosphates, sodium hydroxide, alkali salts
- Phosphates, sodium hydroxide, alkali salts
- pH:
- 12.3 (3 g/l, H2O, 20°C)
- 12.3 (3 g/l, H2O, 20°C)
- 12.3 (3 g/l, H2O, 20°C)
- Solubility:
- (20°C) Soluble
- (20°C) Soluble
- (20°C) Soluble
- Density:
- 900 kg/m3 (Bulk)
- 900 kg/m3 (Bulk)
- 900 kg/m3 (Bulk)