Big Red Apple Swine Enrichment, Bio-Serv
# de catalogue CA89067-880
Fournisseur: Bio-Serv
Spécifications
- Length:22.9 cm (9")
- Color:Red
- Description:Big Red Apple
- Height:22.9 cm (9")
- Cat. no.:CA89067-880
- Supplier No.:K3263
Spécifications
A propos de cet article
Made of polyethylene
Apple scented. Best swine enrichment available. Allows for natural rooting behavior. Hanging loop allows for grab and play. Cagewasher safe.
Customers Who Bought This Also Bought