Bochem Anodized Lab Support Jacks with Stainless Steel Lifts, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
Improved–Jacks now feature thicker scissor mechanism supports for enhanced stability.
- 304 stainless lift mechanisms for long service life
- Anodized aluminum decks
Lift mechanism is constructed entirely of 304 stainless steel with POM rollers and phenolic knob for long service life in corrosive environments, such as fume hoods. All-roller design provides smooth stable height adjustment. Large (30x30cm and 40x40cm) sizes are supplied with a ratcheting wrench for easy height adjustment. Jacks are manufactured according to DIN 12897.
Ordering information: Models B11081 and B11091 include ratchet to assist in adjustment.
Customers Who Bought This Also Bought