Bochem Powder Coated Lab Support Jacks with Stainless Steel Lifts, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
Improved–Jacks now feature thicker scissor mechanism supports for enhanced stability.
- 304 stainless lift mechanisms for long service life
- Epoxy-powder coated aluminum decks
Lift mechanism is constructed entirely of 304 stainless steel with POM rollers and phenolic knob for long service life in corrosive environments, such as fume hoods. All-roller design provides smooth stable height adjustment. Large (30x30cm and 40 x 40 cm) sizes are supplied with a ratcheting wrench for easy height adjustment. Jacks are manufactured according to DIN 12897.
Ordering information: Models B11080 and B11090 include ratchet to assist in adjustment.
Customers Who Bought This Also Bought