ContactUsLinkComponent
Prostate Biopsy Collection and Transport Kits, Therapak®
Prostate Biopsy Collection and Transport Kits, Therapak®
# de catalogue 10497-194
Fournisseur:  Therapak, LLC
ProSterile biopsy kit 8vlpk 30pkcs
Prostate Biopsy Collection and Transport Kits, Therapak®
# de catalogue 10497-194
Fournisseur:  Therapak, LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    8-Vial Prostate Biopsy Transport Kit
  • Supplier No.:
    38869
Spécifications
A propos de cet article
All kits feature standardized components to maximize the ease of collecting and packing specimens for transport.
The kits come with 20 ml jars pre-filled with 10 ml of formalin solution. Jars are secured by a corrugated insert and placed into the provided zip-style biohazard bag with absorbent material for transport.
Corrugated inserts are printed with the prostate biopsy location ID for easy recognition. In addition, a set of jar labels with the location ID is included in the kit to ensure proper placement of the specimen jars at the point of packing and identification once received at the test location.
Primary receptacles in the form of 20 ml jars are 95 kPa certified for transport in compliance with DOT and IATA standards for leak resistant transportation.
Customers Who Bought This Also Bought