Zip Style Reclosable Transport Bags, Two-Pouch with BioHazard Symbol, Therapak®
Fournisseur: Therapak, LLC
Certificats
A propos de cet article
Therapak® zip style bags are clear and feature a biohazard symbol imprinted on the outside.
- Re-sealable
- Conveniently holds specimen and paperwork
The two-pouch bag is manufactured to hold specimen in one pouch and a requisition form in the other.
Ordering information: Bags are also available without the biohazard symbol.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.