About this item
Therapak® assembles Non-DOT, DOT and SAMHSA compliant collection and transport kits using the popular 45 mL capacity tamper evident vial. The vial is constructed of virgin polypropylene that results in excellent clarity for visibility into the collected specimen container. Each vial is graduated at the 15 mL and 30 mL capacity level. Kit configurations include a two-pouch specimen bag with absorbent, a 180 mL beaker style collection cup with pour spout and either one or two 45 mL vials depending on whether your requirements call for single or split collection kits. Kit components are heat-sealed in a bag to protect the integrity of the components. Individual box kits are also listed below.
- Features wide-mouth collection cup
- Includes 45 mL tamper-evident vial
- Single and split kit configurations
- Bag and box kits available
- Customization available upon request