GsBP®-XLB Non-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: PAH, PCB, Benzene, Dioxane.
- Proprietary phase with higher phenyl content than GsBP-5
- Equivalent to USP G41
Ordering information: These columns are suitable replacements for DB-XLB, Rxi-XLB, CP-Sil 13CB, VF-XMS.
Spécifications
- Catalog No:
- 10499-420
- 10499-400
- 10499-402
- 10499-404
- 10499-414
- 10499-392
- 10499-394
- 10499-416
- 10499-396
- 10499-418
- 10499-398
- 10499-406
- 10499-410
- 10499-388
- 10499-412
- 10499-390
- Description:
- 10499-420
- 10499-400
- 10499-402
- 10499-404
- 10499-414
- 10499-392
- 10499-394
- 10499-416
- 10499-396
- 10499-418
- 10499-398
- 10499-406
- 10499-410
- 10499-388
- 10499-412
- 10499-390
- Temperature limit:
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C