A propos de cet article
Typical Application: Acidic/neutral drugs, alkaloids, amines and nitriles, amphetamine and methamphetamine, antiepileptic, basic drugs, chlorinated pesticides, EPA method 508, CLP standard, semivolatile organics, diesel fuel, drug of abuse, endocrine disruptors: alkyl phenols, endocrine disruptors: phthalate, EPA 608.1, EPA air analysis method TO-15, EPA method 525.2, EPA method 551.1, EPA method 610, EPA method 8061 (phthalate ester), EPA method 8270, EPA-625 phenols, flavor mixture, food packaging volatiles, formaldehyde, 50ppb, fragrance allergens, gasoline, halogenated compounds, local anesthetics, nitrogen/phosphorus containing pesticides, EPA 507, organochlorine pesticides, organochlorine pesticides II EPA method 8081A, organohalide pesticides in water, EPA 505, organophosphorous pesticides I EPA 8141A, phenols, I and II, polybrominated diphenyl esters (PBDE), polyethyleneamines, polynuclear aromatic hydrocarbons (PAHs), semivolatile compounds, semivolatile organics, substituted anilines, sulfur in air, trace active amines, 10 ng on-column, tricyclic antipsychotics, urine drug screen, US EOA method 8270D mix.
- 5% phenyl 95% methyl polysiloxane phase
- Equivalent to USP G27, G41, G66
Spécifications
- Catalog No:
- 10499-138
- 10499-144
- 10499-146
- 10499-148
- 10499-150
- 10499-140
- 10499-142
- 10499-196
- 10499-198
- 10510-888
- 10499-380
- 10499-382
- 10499-200
- 10499-384
- 10499-210
- 10499-212
- 10499-206
- 10499-208
- 10499-174
- 10499-176
- 10499-178
- 10499-360
- 10499-370
- 10499-282
- 10499-284
- 10499-372
- 10499-286
- 10499-374
- 10499-288
- 10499-376
- 10499-362
- 10499-364
- 10499-366
- 10499-280
- 10499-368
- 10499-188
- 10499-190
- 10499-192
- 10499-194
- 10499-290
- 10499-378
- 10499-180
- 10499-182
- 10499-186
- 10499-152
- 10499-154
- 10499-156
- 10499-158
- 10499-262
- 10499-264
- 10499-266
- 10499-342
- 10499-344
- 10499-346
- 10499-166
- 10499-168
- 10499-278
- 10499-170
- 10499-172
- 10499-268
- 10499-356
- 10499-160
- 10499-270
- 10499-358
- 10499-272
- 10499-162
- 10499-164
- Description:
- 10499-138
- 10499-144
- 10499-146
- 10499-148
- 10499-150
- 10499-140
- 10499-142
- 10499-196
- 10499-198
- 10510-888
- 10499-380
- 10499-382
- 10499-200
- 10499-384
- 10499-210
- 10499-212
- 10499-206
- 10499-208
- 10499-174
- 10499-176
- 10499-178
- 10499-360
- 10499-370
- 10499-282
- 10499-284
- 10499-372
- 10499-286
- 10499-374
- 10499-288
- 10499-376
- 10499-362
- 10499-364
- 10499-366
- 10499-280
- 10499-368
- 10499-188
- 10499-190
- 10499-192
- 10499-194
- 10499-290
- 10499-378
- 10499-180
- 10499-182
- 10499-186
- 10499-152
- 10499-154
- 10499-156
- 10499-158
- 10499-262
- 10499-264
- 10499-266
- 10499-342
- 10499-344
- 10499-346
- 10499-166
- 10499-168
- 10499-278
- 10499-170
- 10499-172
- 10499-268
- 10499-356
- 10499-160
- 10499-270
- 10499-358
- 10499-272
- 10499-162
- 10499-164
- Temperature limit:
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C