A propos de cet article
80-400 gloves provide excellent insulation and comfortable handling in cold conditions.
- Excellent insulation against the cold
- Extra safety due to insulation and high-visibility features
The combination of a thermal terry loop acrylic liner with a naturally insulating latex coating ensures excellent insulation against the cold in temperatures down to as low as 13 °C for short or intermittent contact.
Use in the public utilities, recycling, forestry, general carpentry, general industry, masonry tile, and warehousing industries. Uses include outdoor construction work; cold storage and unheated warehouses; forestry, logging and lumber operations; general maintenance; handling chilled or refrigerated products; and patrolling unheated premises.
The terry loop acrylic liner is bright yellow in color, offering extra high visibility in low or poor light conditions. This extra visibility brings additional safety to external workers when it is getting dark.
Spécifications
- Catalog No:
- 89012-784
- 89012-786
- 89012-790
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Liner material:
- Acrylic/Terry
- Acrylic/Terry
- Acrylic/Terry
- Finish:
- Palm Coated
- Palm Coated
- Palm Coated
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- EN norm:
- X231
- X231
- X231