A propos de cet article
Amines & Phenols, Amphetamines and Precursors - TMS Derivative, Antihistamines, Aroclors 1016-1268 (without 1221), Bacterial Fatty Acid Methyl Esters, Basic Drugs (Underivatized), Benzodiazepines (Basic Drugs - Underivatized), Bergamot Oil, Butter Triglycerides, Cannabinoids (TMS Derivatives), Chlorinated hydrocarbons, CLP Pesticides, Cocaine & Metabolites (TMS Derivatives), Cold Medication, Cold-Pressed Orange Oil, Common Drug Screen, Congeners in DIN Method PCBs, Ethanolamines, Typical Application: Ethyleneamines, European Red List Volatiles, Garlic Oil, Herbicides, Lemon Oil, Marijuana - TMS Derivatives, Major Metabolites - TMS Derivatives, Motor Oil, Narcotics and Adulterants, Opiates (TMS Derivatives), Organic Volatile Impurities, Organochlorine Pesticides, Organomercury Compounds, Organotin Compounds II, Pain Killers - TMS Derivatives, PBDEs, PCBs by EPA Method 8083, Phencyclidine (PCP), Phytosterols (Saw Palmetto), Steroids: Anabolic (Underivatized), Sympathomimetic Amines Derivatives, Ylang Ylang Oil.
- 5% phenyl 95% methyl polysiloxane phase
- Equivalent to USP G27, G41, G66
Spécifications
- Catalog No:
- 10498-896
- 10498-898
- 10498-976
- 10498-888
- 10498-978
- 10498-890
- 10498-892
- 10498-894
- 10498-980
- 10498-902
- 10498-990
- 10498-992
- 10498-904
- 10498-994
- 10498-906
- 10499-060
- 10498-996
- 10499-062
- 10498-908
- 10498-982
- 10498-984
- 10498-986
- 10498-988
- 10498-900
- 10498-874
- 10498-876
- 10498-954
- 10498-956
- 10498-958
- 10499-304
- 10499-306
- 10499-300
- 10499-302
- 10498-968
- 10499-056
- 10498-970
- 10499-058
- 10498-882
- 10498-884
- 10498-972
- 10498-886
- 10498-974
- 10498-960
- 10498-962
- 10498-964
- 10499-316
- 10498-966
- 10499-054
- 10498-932
- 10498-934
- 10498-936
- 10498-938
- 10498-946
- 10498-948
- 10498-952
- 10498-940
- 10498-942
- 10498-944
- 10498-918
- 10499-072
- 10499-074
- 10498-910
- 10499-064
- 10498-998
- 10499-066
- 10498-912
- 10498-914
- 10499-068
- 10498-916
- 10499-070
- 10499-000
- 10499-002
- 10498-924
- 10499-012
- 10498-926
- 10498-928
- 10498-930
- 10499-004
- 10499-006
- 10499-008
- 10498-920
- 10498-922
- 10499-010
- Description:
- 10498-896
- 10498-898
- 10498-976
- 10498-888
- 10498-978
- 10498-890
- 10498-892
- 10498-894
- 10498-980
- 10498-902
- 10498-990
- 10498-992
- 10498-904
- 10498-994
- 10498-906
- 10499-060
- 10498-996
- 10499-062
- 10498-908
- 10498-982
- 10498-984
- 10498-986
- 10498-988
- 10498-900
- 10498-874
- 10498-876
- 10498-954
- 10498-956
- 10498-958
- 10499-304
- 10499-306
- 10499-300
- 10499-302
- 10498-968
- 10499-056
- 10498-970
- 10499-058
- 10498-882
- 10498-884
- 10498-972
- 10498-886
- 10498-974
- 10498-960
- 10498-962
- 10498-964
- 10499-316
- 10498-966
- 10499-054
- 10498-932
- 10498-934
- 10498-936
- 10498-938
- 10498-946
- 10498-948
- 10498-952
- 10498-940
- 10498-942
- 10498-944
- 10498-918
- 10499-072
- 10499-074
- 10498-910
- 10499-064
- 10498-998
- 10499-066
- 10498-912
- 10498-914
- 10499-068
- 10498-916
- 10499-070
- 10499-000
- 10499-002
- 10498-924
- 10499-012
- 10498-926
- 10498-928
- 10498-930
- 10499-004
- 10499-006
- 10499-008
- 10498-920
- 10498-922
- 10499-010
- Temperature limit:
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 300/320°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 300/320°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 260/280°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C