GsBP®-Tubing Deactivated FS GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
GC tubing.
Spécifications
- Catalog No:
- 10500-120
- 10500-122
- 10500-124
- 10500-134
- 10500-136
- 10500-138
- 10500-126
- 10500-128
- 10500-130
- 10500-132
- 10500-140
- 10499-260
- 10500-142
- 10500-144
- 10500-146
- 10499-254
- 10500-112
- 10500-156
- 10500-158
- 10500-114
- 10499-256
- 10500-116
- 10499-258
- 10500-148
- 10500-150
- 10500-108
- 10500-152
- 10500-154
- 10500-110
- Temperature limit:
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 340/360°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325°C
- -60 to 325/350°C
- -60 to 325°C
- -60 to 325°C
- -60 to 325/350°C
- -60 to 340/360°C
- -60 to 325/350°C
- -60 to 325°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C