GsBP®-PLOT-U GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: Amines in Water, CO₂ in air, Hydrocarbon Gases, Moisture, N₂/CO, Primary Amines, Solvents, Sulfurs.
- Divinylbenzene/ethylene acrylate porous co-polymer
- Immobilized
- Polar PLOT phase
- Equivalent to USP G45, S4
Ordering information: These columns are suitable replacements for HP-PLOT U, CP-Porapak U.
Spécifications
- Catalog No:
- 10500-068
- 10500-070
- 10499-246
- 10499-248
- 10499-250
- 10499-252
- Temperature limit:
- -80 to 190/200°C
- -80 to 190/200°C
- -80 to 190/200°C
- -80 to 190/200°C
- -80 to 190/200°C
- -80 to 190/200°C