GsBP®-PLOT-Q GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: Air, Carbon Monoxide, Methane, Carbon Dioxide, Alcohols, Fatty Acids (Free), Gases (Air, Carbon Monoxide, Methane, Carbon Dioxide), Hydrocarbon Gases, N2OI, Natural Gas, Nitrogen, Oxygen, Argon, Oxygenates, Refinery Gas I, Solvents, Sulfur Gas Analysis in Light HydroCarbon Streams.
- Divinylbenzene porous polymer
- Immobilized
- Equivalent to USP S2, S3, S6
Ordering information: These columns are suitable replacements for HP-PLOT Q, CP-Porapak Q, CP-Porabond Q,Rt-Q.
Spécifications
- Catalog No:
- 10500-052
- 10499-722
- 10500-054
- 10500-056
- 10500-058
- 10499-720
- 10499-716
- 10499-718
- 10500-062
- 10500-064
- 10500-066
- Description:
- 10500-052
- 10499-722
- 10500-054
- 10500-056
- 10500-058
- 10499-720
- 10499-716
- 10499-718
- 10500-062
- 10500-064
- 10500-066
- Temperature limit:
- -80 to 280°C
- -80 to 250/260°C
- -80 to 260/280°C
- -80 to 260/280°C
- -80 to 260/280°C
- -80 to 250/260°C
- -80 to 250/260°C
- -80 to 250/260°C
- -80 to 250/260°C
- -80 to 250/260°C
- -80 to 250/260°C