GsBP®-GasPro GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: C1 and C2 halocarbons (freons), extended hydrocarbon analysis, halocarbons, halothane, inorganic gases, ,ercaptans, residue solvents, sulfur compounds in propylene (1 ppm), sulfur gas analysis in light hydrocarbon streams I.
- PLOT phase
- Immobilized
Ordering information: These columns are suitable replacements for GS-GasPro, CP-Select Silica.
Spécifications
- Catalog No:
- 10500-046
- 10500-048
- 10500-050
- Description:
- 10500-046
- 10500-048
- 10500-050
- Temperature limit:
- -80 to 260/300°C
- -80 to 260/300°C
- -80 to 260/300°C