GsBP®-PLOT-MS5A GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: Air, Argon, CO, Hydrogen, Natural gas, Nitrogen Oxides, Noble gases, Refinery has, SF6.
- PLOT phase
- Molecular sieve 5A
- Immobilized
Ordering information: These columns are suitable replacements for HP-PLOT Molsieve, CP-PLOT Molesieve.
Spécifications
- Catalog No:
- 10500-032
- 10500-034
- 10500-044
- 10500-040
- 10500-042
- 10500-038
- Description:
- 10500-032
- 10500-034
- 10500-044
- 10500-040
- 10500-042
- 10500-038
- Temperature limit:
- -80 to 300°C
- -80 to 300°C
- -80 to 300°C
- -80 to 300°C
- -80 to 300°C
- -80 to 300°C