GsBP®-PLOT-S GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: Refineral gas, Propane, Propylene, Butadienes, CFCs, C1 to C5, Extended C1 toC10, Nitrogen oxides.
- PLOT phase
- Al₃O₃
- Modified by Na₂SO₄
- Immobilized
Ordering information: These columns are suitable replacements for HP-PLOT AL₂O₃, CPAlumina PLOT, Rt-Alumina, GS-Alumina.
Spécifications
- Catalog No:
- 10500-008
- 10500-010
- 10500-012
- 10500-014
- 10500-000
- 10500-002
- 10500-004
- 10500-006
- Description:
- 10500-008
- 10500-010
- 10500-012
- 10500-014
- 10500-000
- 10500-002
- 10500-004
- 10500-006
- Temperature limit:
- -60 to 200°C
- -60 to 200/250°C
- -60 to 200°C
- -60 to 200°C
- -60 to 200/250°C
- -60 to 200/250°C
- -60 to 200/250°C
- -60 to 200°C