GsBP®-PLOT-KCL GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical application: Refineral gas, Propane, Propylene, Butadienes, CFCs, C1 to C5, Extended C1 toC10, Nitrogen oxides.
- PLOT phase
- Al3O3
- Modified by KCL
- Immobilized
Ordering information: These columns are suitable replacements for HP-PLOT AL2O3, CPAlumina PLOT, Rt-Alumina, GS-Alumina.
Spécifications
- Catalog No:
- 10499-240
- 10499-242
- 10499-992
- 10499-244
- 10499-238
- 10499-994
- 10499-996
- 10499-998
- Description:
- 10499-240
- 10499-242
- 10499-992
- 10499-244
- 10499-238
- 10499-994
- 10499-996
- 10499-998
- Temperature limit:
- -60 to 200°C
- -60 to 200°C
- -60 to 200/250°C
- -60 to 200°C
- -60 to 200°C
- -60 to 200/250°C
- -60 to 200/250°C
- -60 to 200°C