GsBP®-1701MS Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: 5% Fragrance Materials Association Mix, Acrylate Impurities II, Antidepressants (Basic Drugs- Uncategories), Antiepileptics, Barbiturates (Acidic/Neutral Drugs-Underivatized), Fentanyls, Organochlorine Pesticides III, Phenoxy Acid Herbicides, Styrene Impurities, TMS derivatized Sugars, Triazine Herbicides I, VOCs in Packaging materials.
- 14% cyqnopropyl,86% methyl polysiloxane phase
- Mid-polar
Spécifications
- Catalog No:
- 10499-988
- 10499-990
- 10499-982
- 10499-984
- 10510-876
- Description:
- 10499-988
- 10499-990
- 10499-982
- 10499-984
- 10510-876
- Temperature limit:
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C