GsBP®-502.2 Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Equivalent to: DB-624, DB-VRX, Rtx-624, Rtx-VMS, DB 502.2, VOCCol.
- Proprietary phase
- Close to GsBP-1301
- Mid-polar
- USP Code: G43
Spécifications
- Catalog No:
- 10499-942
- 10499-940
- 10499-936
- 10499-938
- 10499-934
- Description:
- 10499-942
- 10499-940
- 10499-936
- 10499-938
- 10499-934
- Temperature limit:
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260/280°C