GsBP®-1301 Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: 1,4-Dioxane, EPA 8010, EPA Method 551.1, Organic Volatile Impurities, Residual Solvents - European Pharmacopoeia Class I,II, Spirit drinking, Rum, Scotch.
- 6% cyqnopropyl,94% methyl polysiloxane phase
- Mid-polar
- Equivalent to USP G43
Ordering information: These columns are suitable replacements for DB-1301, CP-1301, VF-1301, Rtx-1301.
Spécifications
- Catalog No:
- 10499-800
- 10499-788
- 10499-792
- 10499-794
- 10499-802
- 10499-782
- 10499-784
- 10499-786
- 10499-234
- 10499-236
- 10499-796
- 10499-224
- 10499-226
- 10499-798
- 10499-228
- Description:
- 10499-800
- 10499-788
- 10499-792
- 10499-794
- 10499-802
- 10499-782
- 10499-784
- 10499-786
- 10499-234
- 10499-236
- 10499-796
- 10499-224
- 10499-226
- 10499-798
- 10499-228
- Temperature limit:
- -20 to 260/280°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 260/280°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 260/280°C
- -20 to 280/300°C