GsBP®-608 Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Similar phases include DB-608.
Ordering information: These columns are suitable replacements for DB-608.
Spécifications
- Catalog No:
- 10499-222
- 10499-780
- 10499-776
- 10499-778
- Description:
- 10499-222
- 10499-780
- 10499-776
- 10499-778
- Temperature limit:
- 40 to 280/300°C
- 40 to 280/300°C
- 40 to 320/340°C
- 40 to 320/340°C