GsBP®-35MS Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical application: Amytriptyline and Nortriptyline, Anilines, Barbiturates (Acid-neutral drugs), Basic drugs (Underivatized), Benzodiazepines, Chlordane, CLP Pesticides, Cold medication (Basic drugs - Underivatized), Endocrine disruptos: Butyl tins (hexyl derivatives), EPA Method 552.2, EPA Method 615-Chlorophenoxyacid herbicides, Ethanolamines, Fentanyl, Organo Tins, Organochlorine pesticides I EPA Method 8081A, Organochlorine pesticides IV, Organophosphorus pesticides II EPA 8141A, PCBs by EPA Method 8082, Pesticides, EPA 508.1, Phenols, Phenoxy acid herbicides - Methyl Derivative, EPA 8151A, Primary amines, Sympathomimetic amines (Basic drugs - Underivatized), Toxaphene.
- 35% phenyl 65% methyl polysiloxane phase
- Mid-polar
- Equivalent to USP G28, G32 and G42
Ordering information: These columns are suitable replacements for DB-35, DB-35MS, HP-35, Rtx-35, ZB-35,VF-35.
Spécifications
- Catalog No:
- 10499-642
- 10499-620
- 10499-644
- 10499-646
- 10499-624
- 10499-612
- 10499-634
- 10499-636
- 10499-614
- 10499-616
- 10499-618
- 10499-628
- 10499-606
- 10499-630
- 10499-608
- 10499-610
- 10499-632
- Description:
- 10499-642
- 10499-620
- 10499-644
- 10499-646
- 10499-624
- 10499-612
- 10499-634
- 10499-636
- 10499-614
- 10499-616
- 10499-618
- 10499-628
- 10499-606
- 10499-630
- 10499-608
- 10499-610
- 10499-632
- Temperature limit:
- 40 to 300/320°C
- 40 to 320/340°C
- 40 to 280/300°C
- 40 to 280/300°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C
- 40 to 320/340°C