GsBP®-CarboWax-20M Polar, PEG GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
A propos de cet article
Typical Application: Acids, Amines, Alcohols, Wine/Liqueur.
Similar phases include HP-20M, DB-CAM, CP-Sil 57Wax.
Ordering information: These columns are suitable replacements for HP-20M, DB-CAM, CP-Sil 57Wax.
Spécifications
- Catalog No:
- 10499-580
- 10499-590
- 10499-568
- 10499-592
- 10499-594
- 10499-572
- 10499-574
- 10499-582
- 10499-584
- 10499-562
- 10499-586
- 10499-564
- 10499-588
- 10499-566
- 10499-576
- 10499-578
- Description:
- 10499-580
- 10499-590
- 10499-568
- 10499-592
- 10499-594
- 10499-572
- 10499-574
- 10499-582
- 10499-584
- 10499-562
- 10499-586
- 10499-564
- 10499-588
- 10499-566
- 10499-576
- 10499-578
- Temperature limit:
- 40 to 220°C
- 50 to 200°C
- 40 to 220°C
- 50 to 200°C
- 50 to 200°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C
- 50 to 200°C
- 40 to 220°C
- 40 to 220°C
- 40 to 220°C