Marathon MicroDL™ Temperature Data Loggers, Therapak®
Supplier: Therapak, LLC
Certificates
About this item
The Marathon MicroDL™ is a programmable data logger engineered to display realtime temperature that also offers the ability to scroll through a summary of recorded history information
The operating temperature range is –30 to 72 °C (–22 to 162 °F). MicroDL™ Reader and Software allows the data logger to be programmed and provides easy access to temperature graphs and data.
Specifications
- Catalog No:
- 10497-132
- 10497-134
- 10497-128
- Interface:
- USB