List of Contents Card, Therapak®
# de catalogue 10029-362
Fournisseur: Therapak, LLC
Spécifications
- Description:List of contents card
- Cat. no.:10029-362
- Dimensions:10.8×6.9 cm (41/4×23/4")
- Supplier No.:54765
Spécifications
A propos de cet article
IATA Packing Instruction 650 requires "an itemized list of contents" card as part of the packaging instruction.
This card should be placed between the secondary packaging and the outer packaging. The standardized list of contents card is available in any printed quantity requested.
Ordering information: Customization available on request.
Customers Who Bought This Also Bought