Histology Bucket with Lid, Therapak®
Supplier: Therapak, LLC
Certificates
About this item
These polypropylene histology buckets are conveniently packed in case quantities and each bucket features a carrying handle and snap-on lid
The lid secures tightly to the bucket to reduce odor. The buckets are autoclavable, easy to stack, and are resistant to cracking when exposed to common cytology and histology based chemicals such as formaldehyde.
Customers Who Bought This Also Bought