VWR® Bulk Beads
Fournisseur: VWR International
Certificats
A propos de cet article
Bead beating is the most effective and verstile lysis method available.
- Certified free of RNase and DNase
- Designed for use on all bead mill platforms
- Effective and versatile
VWR® offers optimal flexibility with a range of bead beating lysing matrices to support efficient homogenization of a wide range of samples.
VWR® bulk lysing matrix bead media is certified RNase and DNase free to ensure high analyte integrity. Beads are designed for use on all bead mill platforms including the VWR® Mini Bead Mill.
Certifications: RNase- and DNase-free.
Customers Who Bought This Also Bought