VWR® Pre-Filled Bead Tubes, 1.5 ml
Supplier: VWR International
Certificates
About this item
VWR® pre-filled bead tubes offer convenience and flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples on most popular bead mill platforms.
- Certified RNase and DNase free to ensure high analyte integrity
The VWR® 1.5 ml pre-filled bead tubes are compatible with the VWR® 4-Place Mini Bead Mill, Omni International Bead Ruptor 12, and Next Advance Bullet Blender.
These 1.5 ml polypropylene microtubes feature an attached lid and retaining cam. They are pre-filled with lysing matrix. Bead beating tubes are designed to withstand the high forces generated through bead milling.
These VWR® bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high yield sample homogenates.
Ordering information: Unit includes 50 bead beating tubes pre-filled with lysing matrix.
Specifications
- Catalog No:
- 10158-600
- 10158-602
- 10158-598
- Operating temperature:
- –80 to 121°C (–112 to 250°F)
- –80 to 121°C (–112 to 250°F)
- –80 to 121°C (–112 to 250°F)