A propos de cet article
SGE has been making precision chromatography syringes for over 50 years
Features and Benefits–Unique plunger design minimizes bending and seizing.
- Unique plunger design minimizes bending and seizing
SGE has been making precision chromatography syringes for over 50 years
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducing instrument downtime, and minimizing the risk of instrument shutdown during the processing of large batches of samples.
Accuracy and reproducibility is ± 1 % (displaced volume); temperature range is 5-120°C removable needle.
Our range of HPLC autosampler syringes are designed and tested to meet critical autosampler specifications.